Learn More
Thermo Scientific™ GC Tool Kits for Thermo Scientific Instruments
Maintain the performance of your GC instrument with Thermo Scientific GC Tool Kits.
Brand: Thermo Scientific™ 60180-784
10090.92 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Maintain the performance of your GC system with Thermo Scientific™ GC Tool Kits. Wrench sets, a flashlight, brushes, a minidrill set, and other tools are included. Kits are available for Thermo Scientific GC and Agilent™ GC instruments.
Features- Replacements included perform like original instrument parts
- Parts list included for easy reordering
Includes:
Capillary instrument gauge, liner cap removal tool, open-end wrenches in 6 x 7mm, 8 x 10mm and 16 x 17mm sizes.
Specifications
Capillary Tool Kit | |
GC Tool Kit for Thermo Scientific Instruments | |
GC | |
Thermo Scientific GC and Agilent GC Instruments |