Learn More
RETSCH Gasket For Grinding Jar
Brand:  RETSCH 22.085.0008
7994.25 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11934362
  • 8630.00 NOK / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Specifications
For 5mL grinding jar
10 Pack
Gasket
For 400 mixer mill
Stainless Steel, PTFE