Learn More
RETSCH Gasket For Grinding Jar
Brand: RETSCH 22.085.0007
4826.00 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Specifications
For 1.5mL grinding jar | |
10 Pack | |
Gasket |
For 400 mixer mill | |
Stainless Steel, PTFE |