Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Side Hole Tip
Designed to provide improved solvent resistance, greater temperature range and increased operational smoothness.
Brand:  Trajan Scientific and Medical™ 005891
 View more versions of this product
Product Code. 13189215
  • 2015.00 NOK / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
PTFE (Plunger)
0.32 mm
Fixed Needle
85 mm
23 G
0.63 mm
100 μL
Side Hole