Learn More
Saint Gobain Life Sciences Gas Sampling Bag
Use these bags for gas sample analysis. Saint Gobain Life Sciences Gas Sampling Bag feature a cylindrical shape and an ebonite tube and tap.
Brand:  Saint Gobain Life Sciences 5607073
Product Code. 10626395
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Bags are individually packaged
  • Available in packs of ten
  • Foil thickness : 5/10mm
  • Custom sizes and configurations available on request
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gas Sampling Bag
1 L
Rubber
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Saint Gobain Life Sciences Gas Sampling Bag > Volume: 1L