Learn More
Ansell™ Gammex™ PF Powder-Free Latex Surgical Gloves
Provides an optimal combination of safety, based on Ansell's powder-free technology, with paralleled levels of comfort and ease-of-use. Ansell Edmont™ Gammex™ PF Powder-Free Latex Surgical Gloves have a very high tear resistance, with a favorable strength-to-thickness ratio for optimal barrier protection.
Brand: Ansell™ AJ330048075
This item is not returnable.
View return policy
Description
- Micro-rough outer surface provides a secure grip in both wet and dry conditions
- Synthetic inner coating reduces possible contact with latex allergens
- Resistant to alcohol-based hand disinfectants and viral penetration
- Delivers elasticity, sensitivity and comfort plus a natural fit that helps prevent hand fatigue
Specifications
Single-use | |
No | |
Powder Free | |
Straight | |
Tears, alcohol-based hand disinfectants, viral penetration |
Ambidextrous | |
Latex | |
7.5 | |
50 Pairs/Box |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Ansell™ Gammex™ PF Powder-Free Latex Surgical Gloves > Size: 7.5