Learn More
Fisherbrand™ Gamma Radiation Shields
Gamma radiation shields with fixed 15 degree angles and a curved base of various dimensions, are ideal for protection from Gamma radiation.
Brand:  Fisherbrand™ CSR-S1OG
 View more versions of this product
Product Code. 12337177
  • 29010.00 NOK / Each
Only null left
Estimated Shipment: 05-11-2025
Description
  • Supplied in small, medium and large sizes and with curved base 15cm-deep for use with safety trays or flat 30x30cm base for under-the-bench protection
  • A range of angles offers increased maneuverability, while clear optical acrylic aids visualization
  • Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
  • Available in two sizes, with either a flat base to provide bench protection, or a curved base which fits conveniently over safety trays
Specifications
Specifications
Small Fixed 45° Angle
Gamma Radiation Shield
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Gamma Radiation Shields > Angle: Small Fixed 45°