missing translation for 'onlineSavingsMsg'
Learn More

Usbeck Laborgeraete™ Galvanized Steel Tripod

Brand:  Usbeck Laborgeraete™ 5141

 View more versions of this product

Product Code. 10600293

  • 319.93 NOK / Each
Only null left
Estimated Shipment: 18-08-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Galvanized Steel
100 mm
1 Ea.
180 mm
Tripod
290 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Usbeck Laborgeraete™ Galvanized Steel Tripod > Inner Dia.: 100mm; Height: 180mm; Weight: 290g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.