missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 7 Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
1632.00 NOK - 4125.00 NOK
Specifications
Antigen | Galectin 7 |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17674266
|
Invitrogen
MA543813 |
20 μL |
1632.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17664266
|
Invitrogen
MA543812 |
100 μL |
4125.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.Specifications
Galectin 7 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
GAL7; Gal-7; galectin 7; galectin 7B; galectin-7; galectin-7B; HKL-14; keratinocyte lectin 14; lectin galactoside-binding soluble 7; lectin, galactose binding, soluble 7; lectin, galactoside binding soluble 7; lectin, galactoside binding soluble 7B; lectin, galactoside-binding, soluble, 7; lectin, galactoside-binding, soluble, 7B; LGALS7; LGALS7A; LGALS7B; p53-induced gene 1 protein; p53-induced protein 1; PI7; Pig1 | |
LGALS7, LGALS7B | |
IgG1 κ | |
Protein A/G | |
Antibody |
C2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
P47929, P97590 | |
29518, 3963, 653499 | |
Met1-Phe136 (Accession P47929), with N-terminal His Tag | |
Primary | |
-20°C | |
LGALS7 |