Learn More
A&D Weighing HR Analytical Balance Internal Calibration
HR Analytical Balance with C-SHS patented weighing technology, which provides performance and a lean compact design.
Brand: A&D Weighing HR-250AZ
17246.95 NOK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Robust construction, strong protection against dust & water
- Built-in internal calibration at the touch of a button (HR-AZ)
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow space
- Reverse backlit LCD display for easy to read weighing results
- SCF – Statistical calculation function
- Standard RS-232C
- Fast compact SHS sensor, response time of about 2 seconds
- Quick USB interface – no driver installation required (optional)
- LAN – Ethernet interface with WinCT-Plus software (optional)
- Easily removable large breeze break
- Built-in rechargeable NiMH battery pack (optional)
Specifications
GLP/GMP/GCP/ISO | |
252 g | |
0.1 mg | |
HR-250AZ | |
11 VA to the AC adaptor (12 V, 1 A) |
RS-232 C, Quick USB (option), LAN (option) | |
Reverse-backlit LCD | |
Stainless Steel | |
2 sec. |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
- Multiple Manufacturers
Product Content Correction
Product Title
A&D Weighing HR Analytical Balance Internal Calibration > HR Analytical Balance Internal Calibration; Capacity: 252 g x 0.1 mg