missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumarylacetoacetate hydrolase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-16546
This item is not returnable.
View return policy
Description
Fumarylacetoacetate hydrolase Polyclonal specifically detects Fumarylacetoacetate hydrolase in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Fumarylacetoacetate hydrolase | |
Polyclonal | |
Unconjugated | |
PBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
beta-diketonase, EC 3.7.1.2, FAA, fumarylacetoacetase, Fumarylacetoacetate hydrolase, fumarylacetoacetate hydrolase (fumarylacetoacetase) | |
Rabbit | |
46 kDa | |
0.1 mL | |
Lipid and Metabolism | |
2184 | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.95mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin | |
P16930 | |
FAH | |
Recombinant protein encompassing a sequence within the center region of human Fumarylacetoacetate hydrolase. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (88%), Rat (88%), Bovine (90%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only