Learn More
Memmert™ Full-Sight Glass Door for UN75, Max. 250°C
Full-sight glass door constructed of four insulated glass layers.
Brand: Memmert™ UN75B0
This item is not returnable.
View return policy
Description
- Accessory
- Full-sight
- Glass (4 layers)
- Temperature range up to 250°C
Specifications
UN75 Oven | |
250°C (Max.) | |
Full-sight Door | |
Glass |