Learn More
Epredia™ Frosted Microscope Slides
Premium slides manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to be used in routine histology and microbiology applications.
40.10 NOK - 43.85 NOK
Specifications
Edges 90°, Ground
Frosted Yes
Material Soda Lime Glass
Coating Uncoated
Writing Surface Yes
View More Specs
Products 2
Product Code Brand Thickness (Metric) Price Quantity & Availability  
Product Code Brand Thickness (Metric) Price Quantity & Availability  
17264884
View Documents
Epredia™
AG00000211E01MNZ20
0.8 to 1.0 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17274884
View Documents
Epredia™
AG00000213E01MNZ20
1 to 1.2 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
90°, Ground
Soda Lime Glass
Yes
Yes
Uncoated
76 x 26 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Epredia™ Frosted Microscope Slides