Learn More
3M™ Front-Fitting Reusable Full Face Mask
3M™ Reusable Full Face Mask features a panoramic visor that allows for a virtually unrestricted field of vision. Compatible with the 3M™ Filters, DT Series, this Full Face Mask is suitable for a wide range of wearers and applications.
Brand: 3M™ 7100265075
10910.86 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Panoramic visor to allow for a virtually unrestricted field of vision
- Visor is designed to minimise visual distortion
- Soft, highly flexible and conforming Liquid Silicone Rubber faceseal to enhance user comfort
- Broad faceseal, designed to minimise pressure points and provide an effective seal without over-tightening the head harness
- Matt finish TPE inner mask helps to prevent glare on the inside of the visor
- 4/Cs.
Specifications
Full Face Respirator | |
Small | |
FF-601F |
Product Content Correction
Product Title
3M™ Front-Fitting Reusable Full Face Mask > Face Mask; Size: Small; Model: FF-601F