Learn More
Eppendorf™ Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F7
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand:  Eppendorf™ 6001022210
1966.64 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 15763245
  • 2269.04 NOK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
In Stock
Description
Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F740 series (5-compartment, compartment 1-4), 2 in/53 mm, drawer, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 563 x 140 x 231 (22.2 x 5.5 x 9.1 in)- Max. box number per rack: 16 (4 x 4)- Stainless steel- For ULT freezer: Premium U410 (max. 15 racks), Premium U700 (max. 25 racks), HEF™ U410 (max. 15 racks), CryoCube™ F570 series (max. 25 racks), CryoCube F740 series (5-compartment, compartment 1-4)- For ULT freezer: Premium U410 (max. 240 boxes), Premium U700 (max. 400 boxes), HEF™ U410 (max. 240 boxes), CryoCube™ F570 series (max. 400 boxes), CryoCube F740 series (5-compartment, compartment 1-4)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Freezer Rack
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Eppendorf™ Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F7 >