Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 4 in/102 mm
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand:  Eppendorf™ 6001000410
Product Code. 15753225
  • 1370.46 NOK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 4 in/102 mm, side access, stainless steel- Rack for 102 mm/4.0 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 141 x 141 x 741 (5.6 x 5.6 x 29.2 in)- Max. box number per rack: 7 (7 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 40 racks)- For ULT freezer: Innova C580 (max. 224 boxes), Innova C760 (max. 280 boxes), CryoCube™ FC660 series (max. 280 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Freezer Rack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products