Learn More
Eppendorf™ Freezer Rack: CryoCube™ FC660 series, DWP (53 mm)
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand: Eppendorf™ 6001050111
This item is not returnable.
View return policy
Description
Freezer Rack: CryoCube™ FC660 series, locking rod, DWP (53 mm), side access, stainless steel- Rack for DWP (53 mm)- Max. DWP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 140 x 91 x 721 (5.5 x 3.6 x 28.4 in)- Max. DWP number per rack: 13 (13 x 1)- Stainless steel- For ULT freezer: CryoCube™ FC660/ FC660h (max. 66 racks)- For ULT freezer: CryoCube™ FC660/ FC660h (max. 780 DWP)- With locking rod for increased sample safetySpecifications
Freezer Rack |