Learn More
Hanna Instruments™ Free and Total Chlorine Portable Photometer
Brand: Hanna Instruments™ HI96711
This item is not returnable.
View return policy
Specifications
Portable Photometer | |
Free and Total Chlorine | |
193 mm | |
0 to 5mg/L | |
0.36 kg |
±0.03mg/L | |
69 mm | |
Tungsten Lamp | |
0.01mg/L | |
104 mm |
Product Content Correction
Product Title
Hanna Instruments™ Free and Total Chlorine Portable Photometer > Range: 0.00 to 5.00 mg/L