Learn More
VACUUBRAND™ FPM Spear Sealing Kit
Brand: VACUUBRAND™ 20660137
111.15 NOK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
40 x 42 x 5mm | |
FPM | |
Sealing Ring |
For Small Flanges KF | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
14Hours
11Minutes
17Seconds
56New Offer Save Up to 92%
Product Content Correction
Product Title
VACUUBRAND™ FPM Spear Sealing Kit > Dimensions: 40 x 42 x 5mm