Learn More
Poulten Graf™ Fortuna™ Optifix™ Safety Dispenser
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
Brand:  Poulten Graf™ 101 10727
 View more versions of this product
Product Code. 10252371
  • 6690.00 NOK / Each
Only null left
Estimated Shipment: 22-07-2025
Description
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With certificate, individually numbered
  • Glass dispensing cylinder is coated with plastic to provide breakage protection
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
Safety Dispenser
0.4 to 2 mL
PTFE-encased Piston, Sealing Lip
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Poulten Graf™ Fortuna™ Optifix™ Safety Dispenser > Capacity: 2mL; Includes: PTFE-encased Piston, Sealing Lip