missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, 37% w/w, Stabilized with Methanol, Pharmpur™, Scharlab
Click to view available options
Quantity:
5 L
25 L
Unit Size:
25 L
5 L
Description
- Synonyms: Formalin solution, Formol, Methanal solution, Methyl aldehyde solution
- Molecular Formula: CH2O
- Molar Mass: 30.03g/mol
- CAS: 50-00-0
- EIN ECS No.: 200-001-8
- Ignition Temperature: ∼300°C (pure substance)
- Vapor Pressure: 1.3hPa (formaldehyde)
- Acidity: Passes test
Specifications
Specifications
Melting Point | <-15°C |
Density | 1.09g/cm3 |
Boiling Point | 93°C to 96°C |
Flash Point | 62°C |
Ignition Residue | 0.1% max |
Quantity | 5 L |
Residual Solvents | Analysed according to guideline CPMP/ICH/283/95 |
Stabilizer | Approx. 10% methanol |
Formula Weight | 30.03g/mol |
Grade | Pharmpur™ USP |
Show More |
Safety and Handling



Product Identifier
- Formaldehyde, solution 37% w/w, extra pure, Pharmpur, Ph Eur, BP, USP, stabilized with approx. 10%
- Danger
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : formaldehyde, méthanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Formaldehyde, 37% w/w, Stabilized with Methanol, Pharmpur™, Scharlab >
Spot an opportunity for improvement?Share a Content Correction