Learn More
Gilac™ Food Specimen Containers with Lids
Click to view available options
Capacity (Metric):
3 L
8 L
Quantity:
5 Pk.
7 Pk.
Unit Size:
Each
Description
- Pack of food specimen containers.
- Containers especially for storing food specimen samples complying with HACCP catering standards.
- Markings do not wash off, even in the machine.
Specifications
Specifications
| Product Type | HACCP Food Container |
| Material | HDPE |
| Capacity (English) | 0.8 gal. |
| Capacity (Metric) | 3 L |
| Color | Assorted |
| Height (English) | 4.7 in. |
| Height (Metric) | 120 mm |
| Length (English) | 13.8 in. |
| Length (Metric) | 350 mm |
| Quantity | 5 Pk. |
| Show More |
Product Content Correction
Product Title
Gilac™ Food Specimen Containers with Lids >