missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19847-1-AP-20UL
This item is not returnable.
View return policy
Description
FNIP1 encodes a protein that binds to the tumor suppressor protein folliculin and to AMP-activated protein kinase (AMPK). The encoded protein participates in the regulation of cellular metabolism and nutrient sensing by modulating the AMPK and target of rapamycin signaling pathways. This gene has a closely related paralog that encodes a protein with similar binding activities. Both related proteins also associate with the molecular chaperone heat shock protein-90 (Hsp90) and negatively regulate its ATPase activity and facilitate its association with folliculin. Diseases associated with FNIP1 include Birt-Hogg-Dube Syndrome and Pleural Disease.Specifications
FNIP1 | |
Polyclonal | |
Unconjugated | |
FNIP1 | |
FNIP1, KIAA1961 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100362347, 216742, 96459 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FD7, Q8TF40 | |
FNIP1 | |
FNIP1 Fusion Protein Ag13899 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |