missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNDC3A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25240-1-AP-20UL
This item is not returnable.
View return policy
Description
FNDC3A gene ontology annotations related to this gene include cell-cell adhesion; fertilization; Sertoli cell development; spermatid development.Specifications
FNDC3A | |
Polyclonal | |
Unconjugated | |
FNDC3A | |
FNDC3, FNDC3A, HUGO, KIAA0970 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22862, 319448 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BX90, Q9Y2H6 | |
Fndc3a | |
FNDC3A Fusion Protein Ag18736 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction