missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein-PGC1a Coactivator Peptide, 100 μM

1712.00 NOK - 8310.00 NOK

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Label Type Classic Dyes
Shipping Condition Dry Ice
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10746983
View Documents
Invitrogen™
PV4421
100 μL
1712.00 NOK
Each
Please to purchase this item. Need a web account? Register with us today!
10443483
View Documents
Invitrogen™
PV4422
1 mL
8310.00 NOK
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ligand binding to nuclear receptors causes conformational changes in the receptor, resulting in a cascade of events, including dissociation of repressor proteins, association of coactivator proteins, and assembly of pol II and other transcriptional factors for activation of target genes. TR-FRET based assays can be developed using the LanthaScreen™ panel of fluorescein-labeled coregulator peptides to investigate conformational changes of nuclear receptors upon ligand binding, either by determining the affinity of ligand-bound receptor for different coregulator peptides, or by identifying additional agonists or antagonists via displacement or recruitment of a specific coregulator peptide.

In the LanthaScreen™ TR-FRET RXR beta Coactivator Assay, a terbium (Tb)-labeled anti-GST antibody is used to indirectly label GSTtagged RXR beta protein. An agonist (9-cis Retinoic Acid) added to the receptor which, upon ligand binding, causes a conformational change resulting in recruitment of the Fl-D22 coactivator peptide. The binding of the fluorescent coregulator peptide to RXR beta causes an increase in the TR-FRET emission ratio.

Specifications

Specifications

Store in freezer (-5 to -30°C).
Classic Dyes
Dry Ice
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.