missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein-IKK Substrate

Fluorescein-IKK Substrate

Brand:  Invitrogen PV3507

Product Code. 10329082

  • 10980.00 NOK / 1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A fluorescein-labeled peptide substrate that is primarily used with the IkappaB kinase (IKK) family.

For use in combination with the LanthaScreen™ Tb-IkappaBalpha [pSer32] antibody (PV3562) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluorescein- DRHDSGLDSMKDE
1 mg/mL fluorescein-IKK substrate in water. Store at -20°C and protect from light.
1 mg
Peptide-Based Substrate
Serine/Threonine Kinases
FITC (Fluorescein)
Dry Ice
CHUK, GRK5, GRK6, IKBKB, IKBKE
Biochemical terbium phospho-specific activity
Fluorescence
Fluorescein-IKK
Kinase Substrate
1147, 2869, 3551
1864 Da
IKBKE, CHUK, GRK5, GRK6, TBK1, IKBKB
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3031453A Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPV3507
Lot Number32773P Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberPV3507
Lot Number1182340B Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberPV3507
Lot Number1801488A Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberPV3507
Lot Number32773Q Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberPV3507
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fluorescein-IKK Substrate > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.