Learn More
Brady™ Floor Stand
Floor Stand designed to warn for hazardous areas, putting valuable safety information directly at the point of need.
858.00 NOK
Specifications
Height (English) 25.98 in.
Height (Metric) 660 mm
Shape Rectangle
View More Specs
Products 3
Product Code Brand Displayed Text Price Quantity & Availability  
Product Code Brand Displayed Text Price Quantity & Availability  
11712005
View Documents
Brady™
241941
ENTREE INTERDITE
858.00 NOK
Each
Only null left
Estimated Shipment: 10-07-2025
Add to basket
11796655
View Documents
Brady™
256262
ATTENTION SOL GLISSANT
858.00 NOK
Each
11706665
View Documents
Brady™
256281
ATTENTION ZONE DE TRAVAIL
858.00 NOK
Each
Only null left
Estimated Shipment: 10-07-2025
Add to basket
Description
Description
  • These floor signs are manufactured from polyester, with a clear matt anti-slip laminate to prevent falling accidents while protecting the graphics underneath.
Specifications
Specifications
25.98 in.
660 mm
Rectangle
Documents
Product Certifications
Product Content Correction
Product Title
Brady™ Floor Stand