missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Flat Stainless Steel Cover With 8 Opening 107 mm And Concentric Ring Sets, For Wnb45

Flat stainless steel cover with concentric ring sets for ring set openings: 8 openings / 107 mm diameter

Brand:  Memmert™ W45B02684

Product Code. 16304717

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

29 L and 45 L Baths
Stainless Steel
8
3 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers