Learn More
Ohaus™ Flat Foam Insert
OHAUS™ Flat Foam Insert is suitable for use with vortex mixers.
Brand: Ohaus™ 30400234
248.82 NOK valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.90000kg
Description
- Offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Foam Insert | |
Foam | |
Flat Foam Insert, Pack of 2 | |
2/Pk. |