Learn More
SCAT™ Flat Flange Lids for Glass Reactors
The flat flange lid is usually placed between two flanges to be joined and then tightened with bolts to create a secure connection. Suitable for glass reactors with a SCHOTT™ flat flange.
Brand: SCAT™ 130101
2052.28 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
- Flat flange allows the lid to be securely attached to the reactor body and ensures a reliable seal to prevent leakage
- Compatible according to DIN 12214 (laboratory flat flange) and compatible with glass reactors for seamless integration
- Effortless and hassle-free to assemble, saving time and effort
- Flexibly expand our products to fit your specific needs and meet future needs
- Made entirely of PTFE, for universal chemical resistance to a wide range of harsh chemicals, increasing application versatility and reliability
- Equipped with practical attachments that allow easy connection and use of existing equipment that has ground or GL threads
- Important component for the safe and reliable connection of pipes and vessels in various industrial applications to prevent leaks and keep equipment running smoothly
Specifications
Flat Flange | |
Glass Reactors | |
DN 100 |
Connection of Pipes and Vessels, Preventing Leaks | |
1 x NPT 1/2 in. (f) Centered, 2 x NPT 1/2 in. (f) Directly in Center, 1 x NPT 1/2 in. (f) Beside Center, 1 x NPT 3/4 in. (f) Beside Center, 1 x NPT 1/4 in. (f) Vertical | |
PTFE |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now