Learn More
Fisherbrand™ Flask Clamp for Shaking Incubators
Fisherbrand™ Flask Clamp is designed to be used with a shaking incubator and can hold a variety of flask sizes.
Brand:  Fisherbrand™ H1000-MR-500
 View more versions of this product
Product Code. 16333236
  • 510.64 NOK / Each
The Academic Best Buy Book 2025
Use promo code 25081 at checkout
View Available Promotions
Only null left
Estimated Shipment: 05-09-2025
Add to basket
Description
Description
  • Constructed of stainless steel
  • For use with shaking incubators
  • Designed to hold flasks
Specifications
Specifications
500 ml Flasks
Stainless Steel
1 Ea.
Flask Clamp
16313226
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Flask Clamp for Shaking Incubators > Holds: 500 mL Flasks