Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
Brand: Thermo Scientific™ 36520051
2125.79 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Beschreibung
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
- Manual syringes in industry-standard size
- White scale backing for easy reading
- Solvent-resistant markings
Spezifikation
Use when performing PTV/LVI injections with a dedicated liner for thermal labile compounds.; 36520051 | |
50 mm | |
Reusable | |
250 ÎĽL | |
Side Hole | |
Removable |
23 G | |
Gas Tight GC Autosampler Syringe, Removable Needle | |
Ea. | |
50 mm | |
TriPlus, AS2000, AS200/AS800 Autosampler | |
36550040 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.