Learn More
Syngene Fixed High Intensity Multiplexing LEDs with Peak Band Passes
HILED modules provide imaging performance for fluorescence applications. There are six options that range from UV to IR light. Each module has a narrow band pass filter to minimise cross-talk ideal for multiplexing with fluorescent gels or blots.
Brand: Syngene HILED-B
This item is not returnable.
View return policy
Description
- HILED modules provide imaging performance for fluorescence applications.
- Choose from six options ranging from UV to IR light.
- Each module has a narrow band pass filter to minimise cross-talk between different wavelengths making it ideal for multiplexing with fluorescent gels or blots.
- Syngene GBox chemi systems are all compatible with these modules.
- The new HILED modules offer reduced exposure time, an extended LED life, and a 20 x 20 cm viewing area.
Specifications
Syngene Gel Doc | |
Imaging DNA Gels and Multiplexing | |
Blue | |
HILEDS in Blue |
Product Content Correction
Product Title
Syngene Fixed High Intensity Multiplexing LEDs with Peak Band Passes > HILEDS in Blue