Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Plastic Lid
For use with Centrifuge 5430 and Centrifuge 5430 R
Brand:  Eppendorf™ 5427755004
7250.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 13216938
  • 7279.00 NOK / Each
Description
  • Eppendorf QuickLock rotors close with only 1/4 of a turn
  • Saves time during every day use
  • Takes repetitive stress off your wrist
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Model: F-45-48-11, Includes Lid
48 x 1.5/2 mL
12,700 rpm
Fixed
48
Fixed Angle Rotor
18,210 x g
5430/5430R Centrifuges
Basic plastic lid
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Fixed Angle QuickLock™ Rotor with Plastic Lid > Model: F-45-48-11, Includes Lid