Learn More
Eppendorf™ Fixed Angle Rotors for Concentrator Plus Centrfuge
For use with Concentrator Plus Centrifuge
Brand: Eppendorf™ 5490034007
5390.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Eppendorf Concentrator plus was designed with the customer in mind to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench.
Specifications
Model: F-45-72-8 | |
72 x 0.5 mL | |
Fixed | |
72 |
Fixed Angle Rotor | |
Concentrator Plus Centrifuge | |
Without Lid |
Product Content Correction
Product Title
Eppendorf™ Fixed Angle Rotors for Concentrator Plus Centrfuge > Capacity: 72 x 0.5mL reaction vessels