Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid
For use with Centrifuge 5430 and Centrifuge 5430 R
Brand: Eppendorf™ 5427739009
11731.74 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- Aerosol-tight Eppendorf QuickLock rotors close with only 1/4 of a turn
- Saves time during every day use
- Takes repetitive stress off your wrist
Specifications
Model F-35-6-30 | |
6 x 15/50 mL | |
Fixed | |
6 |
Fixed Angle Rotor | |
5430/5430R Centrifuges | |
Lid |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Product Content Correction
Product Title
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid > Model F-35-6-30, includes lid