Learn More
Mettler Toledo™ Electrode Arm for FiveEasy™ Plus/FiveEasy Meters
Electrode holder allows you to secure sensor when not in use. For use with FiveEasy Plus/FiveEasy pH or conductivity meters.
Brand: Mettler Toledo™ 30239139
811.70 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Includes: Sensor holder and a pole
Description
- Accessory for use with FiveEasy Plus/FiveEasy Meters
- Secure your sensor with this ambidextrous electrode holder
- Attach this on the right or left side of the FiveEasy meter for convenient measurement and to secure sensor when not in use
Specifications
FiveEasy/Plus electrode arm | |
Electrode Arm | |
Sensor holder and a pole | |
FiveEasy, FiveEasy Plus Meter |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
16Hours
22Minutes
35Seconds
37 -
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
57Hours
22Minutes
35Seconds
37