Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Brand:  SCAT™ 107905
355.78 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 14737170
  • 436.00 NOK / Pack of 10
Only null left
Estimated Shipment: 18-08-2025
Description
SCAT fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Specifications
Ferrule
0.06 in.
Ferrule, 10 Pk.
PTFE
White
1.6 mm
Capillary
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SCAT™ Fittings for HPLC Systems > Fittings; Color: White; For Use With: ; Includes: Ferrule, 10 Pk.; Type: Ferrule