Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Brand:  Fisherbrand™ 1003283
 View more versions of this product
Product Code. 11846244
  • 444.00 NOK / Each
Only null left
Estimated Shipment: 11-07-2025
Add to basket
Description
Description
  • Easy-to-open and close cover
  • Muffles noise level
  • Designed to be used as drip basin when flipped over
  • Chemical-resistant plastic
  • Bring your ultrasonic cleaner up to temperature more quickly
  • Contain and prevent spills and leakage
Specifications
Specifications
Lid
Ultrasonic Cleaner, 6.9 and 9.4 L
Medical Laboratories and in Industrial Production Processes
Plastic
Videos
  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners > For use with FB11205 Ultrasonic Unit