Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Brand:  Fisherbrand™ 1003278
 View more versions of this product
Product Code. 10060892
  • 175.00 NOK / Each
Only null left
Estimated Shipment: 16-07-2025
Add to basket
Description
Description
  • Newly developed
  • Reduces noise and accelerates the heating-up process
  • When turned over, cover can be used as a drip-off basin
  • Collected cleaning liquid can be emptied back into the tank
Specifications
Specifications
Lid
Ultrasonic Cleaner, 0.8 L
Plastic
Videos
  • Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners > For S10/S10H Ultrasonic Cleaner