Learn More
Fisherbrand™ SureOne™ Reload Empty Racks
GREENER_CHOICE
For use with Fisherbrand SureOne universal pipet tips
Brand:  Fisherbrand™ T1000ER-FIS
152.48 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
 View more versions of this product
Product Code. 11993456
  • 232.96 NOK / Pack of 10
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Estimated Shipment: 26-06-2025
Add to basket
Description
Description
  • Inserts (not included) sit flush with rack, allowing easy multichannel pipetting
  • Notched clasp designed to prop lid open for autoclave venting
  • Sure stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe lock latch allows for one-handed opening and closing ability—won't pop open when dropped
  • Sure snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Order Info
Inserts not included; must be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Empty Rack
Pipette Tips
1250 ÎĽL
Videos
  • Talking Points - Fisherbrand Elite Pipettors and SureOne Pipettor Tips (EN)
    Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Talking Points - Fisherbrand Elite Pipettors
    SDS
    Documents Certificates
    View accessory products
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Fisherbrand™ SureOne™ Reload Empty Racks > For tip volume: 1250uL