Learn More
Fisherbrand™ Polypropylene Graduated Container Lids
For polypropylene graduated containers 14-955-101,14-955-103, 14-955-104 and 14-955-106
Brand: Fisherbrand™ 14955107
Description
For use with the following Fisherbrand Specimen Containers:
- 14-955-101, -103 (4.5 oz.)
- 14-955-104, -106 (8 oz.)
Specifications
Lid | |
Sterile | |
Polypropylene graduated containers |
Polyethylene | |
Not Autoclavable | |
500/Pk. |
Product Content Correction
Product Title
Fisherbrand™ Polypropylene Graduated Container Lids > Sterile