Learn More
Fisherbrand™ Permanent Markers
Write on all surfaces
Brand:  Fisherbrand™ 90023-45
 View more versions of this product
Product Code. 11774386
  • 164.32 NOK / Pack of 6
Safety Monthly Promotion
Use promo code 24261 at checkout
View Available Promotions
Only null left
Estimated Shipment: 03-07-2025
Add to basket
Description
Description
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
  • Produce precise, ultrathin lines 0.3mm wide
  • Waterproof and fadeproof, permanent ink dries almost instantly
  • An airtight ring seal cap prevents dry-out and ensures instant writing even after lengthy storage
  • Unique stainless-steel long leg allows precise writing in a confined space
Order Info
Assorted includes six opaque black, two vivid red, two blue, and two green pens
Specifications
Specifications
0.3 mm Ultra Fine
Green
Perfect marking pens for lab samples, microscope slides, photographic slides, glass and plastic beakers, and plastic sample bags
0.3 mm
Permanent Pen
Stainless Steel
6 Pk.
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Discover exclusive monthly deals on essential safety products to enhance your lab's protection and compliance.
    Promo Code: 24261
    Expires2025-06-30
Product Content Correction
Product Title
Fisherbrand™ Permanent Markers > Green