Learn More
Fisherbrand™ Permanent Markers
154.40 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
- Produce precise, ultrathin lines 0.3mm wide
- Waterproof and fadeproof, permanent ink dries almost instantly
- An airtight ring seal cap prevents dry-out and ensures instant writing even after lengthy storage
- Unique stainless-steel long leg allows precise writing in a confined space
Order Info
Assorted includes six opaque black, two vivid red, two blue, and two green pens
Specifications
0.3 mm Ultra Fine | |
Black | |
Stainless Steel | |
6 Pk. |
Permanent Pen | |
Not chemical resistant | |
Perfect marking pens for lab samples, microscope slides, photographic slides, glass and plastic beakers, and plastic sample bags | |
0.3 mm |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now