Learn More
Fisherbrand™ Instant Sealing Sterilization Pouches
No tape or heat sealing needed
Brand:  Fisherbrand™
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 12387209
  • 471.32 NOK / Pack of 100
Estimated Shipment: 19-09-2024
Description
No tape or heat sealing needed
Integral sealing strip eliminates the need to tape or heat seal pouches. Simply peel off protective strip, fold along perforation and press firmly to seal pouch.
  • Triple heat-processed seams provide excellent tear and puncture resistance
  • Suitable for steam or EtO gas sterilization
  • Indicator arrows change color when sterilization cycle is complete
  • Made of paper/plastic; medical-grade paper side guards against puncture
  • 2mil-thick blue transparent polyester/polypropylene laminate film side allows contents to be identified quickly and shows any compromise in the film
  • Corner tack seals prevent contamination in the seal area
  • Thumb notch makes opening pouch easy
Specifications
Specifications
Non-sterile
12 in.
18 in.
2 mil
30 cm
46 cm