Description
Kit provides four adjustable-volume pipettes and a stand.
- Four popular pipette sizes (0.2-2μL, 2-20μL, 20-200μL, 100-1000μL) and stand
- Compatible with universal fit pipet tips
Specifications
1 | |
Pipette Kit | |
Four pipette sizes (0.2-2uL, 2-20uL, 20-200uL, 100-1000uL) and stand |
Elite | |
0.2-2 μL, 2-20 μL, 20-200 μL, 100-1000 μL |
Fisherbrand Liquid Handling Update-2020 (00A)
File size: 3.2 MB
Water analysis
File size: 7.3 MB
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Product Content Correction
Product Title
Fisherbrand™ Elite Pipette Kit > Pipette Kit