Learn More
Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
1090.00 NOK - 1115.00 NOK
Specifications
Sterility | Sterile |
---|---|
Type | Electroporation Cuvet |
Material | Polycarbonate |
Includes | Imbedded aluminum electrodes |
For Use With (Equipment) | Electroporator |
Product Code | Brand | Capacity (Metric) | Width (Metric) Gap | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Width (Metric) Gap | Price | Quantity & Availability | |||||
15532423
|
Fisherbrand™
450150 |
90 ÎĽL | 1 mm |
1115.00 NOK
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
15542423
|
Fisherbrand™
450151 |
400 ÎĽL | 2 mm |
1090.00 NOK
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
15552423
|
Fisherbrand™
450152 |
800 ÎĽL | 4 mm |
1115.00 NOK
Pack of 50 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||||
Description
Disposable, color-coded cuvets are compatible with most electroporators.
- Clear, medical grade polycarbonate for high visibility
- Polished aluminum lowers arcing frequency
- Round cap allows single-handed removal
- FB102 has imbedded aluminum electrodes
Specifications
Sterile | |
Polycarbonate | |
Electroporator |
Electroporation Cuvet | |
Imbedded aluminum electrodes |
Product Title
Fisherbrand™ Electroporation Cuvettes Plus™