Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
Brand:  Fisherbrand™ MP-10D
 View more versions of this product
Product Code. 11943015
  • 480.00 NOK / Each
Only null left
Estimated Shipment: 02-09-2025
Specifications
Owl P8 Series Electrophoresis System
10 x 10 cm
0.15 cm
5.9 in.
15 cm
3.9 in.
13 cm
0.57 cm
13 cm
Comb
10
134 ÎĽL
3.9 in.
10 cm
0.22 in.
10 cm
5.11 in.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System > Comb; 10-teeth; 1.5mm thick