Learn More
Fisherbrand™ Vortex Mixer Head Cover
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™ 80863000
299.45 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15766949
  • 391.00 NOK / Each
Only null left
Estimated Shipment: 10-09-2025
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Head Cover
Vortex Mixer
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Vortex Mixer Head Cover > Head Cover