Learn More
Fisherbrand™ Heavy-Gauge Nickel Combustion Boat
Constructed of high-purity nickel and is most popular in iron and steel labs.
Brand:  Fisherbrand™ 11-474-210
Product Code. 11391443
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Constructed of high-purity nickel and is most popular in iron and steel labs.
Most popular in iron and steel labs
  • Constructed of high-purity nickel
Specifications
Specifications
Combustion Boat
Scientific tools used in experiments because of their resistance to heat and caustic chemicals
9.5 mm
89 mm
0.62 in.
3.5 x 0.62 x 0.37 in. (89 x 16 x 9.5 mm)
0.37 in.
3.5 in.
Nickel
16 mm