Learn More
Fisherbrand™ Heavy-Gauge Nickel Combustion Boat
Constructed of high-purity nickel and is most popular in iron and steel labs.
Brand: Fisherbrand™ 11-474-210
This item is not returnable.
View return policy
Description
Constructed of high-purity nickel and is most popular in iron and steel labs.
Most popular in iron and steel labs
- Constructed of high-purity nickel
Specifications
Combustion Boat | |
Scientific tools used in experiments because of their resistance to heat and caustic chemicals | |
9.5 mm | |
89 mm | |
0.62 in. |
3.5 x 0.62 x 0.37 in. (89 x 16 x 9.5 mm) | |
0.37 in. | |
3.5 in. | |
Nickel | |
16 mm |